Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180740 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNRNP70 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SNRNP70 - small nuclear ribonucleoprotein U1 subunit 70 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301069.1 | 24 | UTR 5 | NP_001287998.1 | |||
NM_003089.5 | 24 | UTR 5 | NP_003080.2 | |||
XM_005259178.1 | 24 | Intron | XP_005259235.1 | |||
XM_011527240.2 | 24 | Intron | XP_011525542.2 | |||
XM_011527241.2 | 24 | Intron | XP_011525543.1 |
Set Membership: |
HapMap Validated |