Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611740 MIM: 600953 MIM: 605791 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
BCO2 - beta-carotene oxygenase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IL18 - interleukin 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEX12 - testis expressed 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031275.4 | 168 | Silent Mutation | AGA,AGG | R,R 12 | NP_112565.1 |