Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606048 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MBOAT7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MBOAT7 - membrane bound O-acyltransferase domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146056.2 | 2310 | UTR 3 | NP_001139528.1 | |||
NM_001146082.2 | 2310 | Intron | NP_001139554.1 | |||
NM_001146083.2 | 2310 | UTR 3 | NP_001139555.1 | |||
NM_024298.4 | 2310 | UTR 3 | NP_077274.3 | |||
XM_011527299.2 | 2310 | UTR 3 | XP_011525601.1 | |||
XM_011527300.2 | 2310 | UTR 3 | XP_011525602.1 | |||
XM_017027296.1 | 2310 | Intron | XP_016882785.1 |
TMC4 - transmembrane channel like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145303.2 | 2310 | Intron | NP_001138775.2 | |||
NM_144686.3 | 2310 | Intron | NP_653287.2 | |||
XM_011526486.2 | 2310 | Intron | XP_011524788.1 |
Set Membership: |
HapMap |