Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTAAGAGGGTATGTAATAGTCTCA[A/C]CTGTACTTTGGATAAAGAAAATAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608699 | ||||||||||||||||||||
Literature Links: |
BMPER PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BMPER - BMP binding endothelial regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133468.4 | Intron | NP_597725.1 | ||||
XM_005249633.2 | Intron | XP_005249690.1 | ||||
XM_017011800.1 | Intron | XP_016867289.1 | ||||
XM_017011801.1 | Intron | XP_016867290.1 |