Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGACTCTACGTGGATCAGTATTT[A/G]TATCACTACTGTGTTGTGGATCCCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602876 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OCLN PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
OCLN - occludin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205254.1 | 1135 | Silent Mutation | TTA,TTG | L,L 233 | NP_001192183.1 | |
NM_001205255.1 | 1135 | Intron | NP_001192184.1 | |||
NM_002538.3 | 1135 | Silent Mutation | TTA,TTG | L,L 233 | NP_002529.1 | |
XM_017008913.1 | 1135 | Silent Mutation | TTA,TTG | L,L 233 | XP_016864402.1 | |
XM_017008914.1 | 1135 | Silent Mutation | TTA,TTG | L,L 233 | XP_016864403.1 |
Set Membership: |
HapMap |