Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGAAGAGGGCAGGGAATGAGGCG[A/G]CAGCTGTGCAGGTGCAGATATTTGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607878 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NNT PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NNT - nicotinamide nucleotide transhydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012343.3 | Intron | NP_036475.3 | ||||
NM_182977.2 | Intron | NP_892022.2 | ||||
XM_005248274.4 | Intron | XP_005248331.1 | ||||
XM_005248275.4 | Intron | XP_005248332.1 | ||||
XM_006714461.3 | Intron | XP_006714524.1 | ||||
XM_011514001.2 | Intron | XP_011512303.1 | ||||
XM_011514002.2 | Intron | XP_011512304.1 | ||||
XM_017009293.1 | Intron | XP_016864782.1 |
NNT-AS1 - NNT antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |