Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600618 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ETV6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ETV6 - ETS variant 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001987.4 | Intron | NP_001978.1 | ||||
XM_011520607.2 | Intron | XP_011518909.1 | ||||
XM_011520608.2 | Intron | XP_011518910.1 | ||||
XM_011520609.2 | Intron | XP_011518911.1 | ||||
XM_011520611.2 | Intron | XP_011518913.1 | ||||
XM_011520612.2 | Intron | XP_011518914.1 | ||||
XM_017018990.1 | Intron | XP_016874479.1 | ||||
XM_017018991.1 | Intron | XP_016874480.1 |
Set Membership: |
HapMap Validated |