Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300298 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UPF3B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
UPF3B - UPF3 regulator of nonsense transcripts homolog B (yeast) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023010.3 | Intron | NP_075386.1 | ||||
NM_080632.2 | Intron | NP_542199.1 | ||||
XM_017029737.1 | Intron | XP_016885226.1 | ||||
XM_017029738.1 | Intron | XP_016885227.1 | ||||
XM_017029739.1 | Intron | XP_016885228.1 | ||||
XM_017029740.1 | Intron | XP_016885229.1 |