Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGAGACAGGAAATGCAGTTTATT[C/T]TAGGTGTCCTTGTGCTCAGCTGAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607898 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIB3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TRIB3 - tribbles pseudokinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301188.1 | Intron | NP_001288117.1 | ||||
NM_001301190.1 | Intron | NP_001288119.1 | ||||
NM_001301193.1 | Intron | NP_001288122.1 | ||||
NM_001301196.1 | Intron | NP_001288125.1 | ||||
NM_001301201.1 | Intron | NP_001288130.1 | ||||
NM_021158.4 | Intron | NP_066981.2 | ||||
XM_017027989.1 | Intron | XP_016883478.1 |
Set Membership: |
HapMap |