Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTTACTATTTGGAGAGTTTCAC[A/G]TATGTAACAATTATTTTGCTGTAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607886 MIM: 607887 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CMTM3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CMTM3 - CKLF like MARVEL transmembrane domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144601.3 | Intron | NP_653202.1 | ||||
NM_181553.2 | Intron | NP_853531.1 | ||||
XM_006721131.1 | Intron | XP_006721194.1 | ||||
XM_011522861.1 | Intron | XP_011521163.1 |
CMTM4 - CKLF like MARVEL transmembrane domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178818.2 | Intron | NP_848933.1 | ||||
NM_181521.2 | Intron | NP_852662.1 | ||||
XM_011522882.1 | Intron | XP_011521184.1 | ||||
XM_017022954.1 | Intron | XP_016878443.1 |
Set Membership: |
HapMap |