Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607170 MIM: 610925 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRELD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CRELD1 - cysteine rich with EGF like domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031717.3 | Intron | NP_001026887.1 | ||||
NM_001077415.2 | Intron | NP_001070883.1 | ||||
NM_015513.4 | Intron | NP_056328.2 | ||||
XM_011534108.1 | Intron | XP_011532410.1 | ||||
XM_017007175.1 | Intron | XP_016862664.1 |
IL17RC - interleukin 17 receptor C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRRT3 - proline rich transmembrane protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |