Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGCTTCAGCCAGTGTGTGTGGAA[A/G]AAACGCCGGTTTTGGGATACGACGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603302 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADCY9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ADCY9 - adenylate cyclase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001116.3 | Intron | NP_001107.2 | ||||
XM_005255079.3 | Intron | XP_005255136.1 | ||||
XM_011522353.2 | Intron | XP_011520655.1 |
LOC102724927 - uncharacterized LOC102724927 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |