Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAGAAACCAACACTCTCTCAGGA[A/C]CAAGCTGGATTCTTCTGAAAGTCGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164017 MIM: 601490 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNRNPA1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
HNRNPA1 - heterogeneous nuclear ribonucleoprotein A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NFE2 - nuclear factor, erythroid 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136023.2 | Intron | NP_001129495.1 | ||||
NM_001261461.1 | Intron | NP_001248390.1 | ||||
NM_006163.2 | Intron | NP_006154.1 | ||||
XM_005268906.4 | Intron | XP_005268963.1 |
Set Membership: |
HapMap Validated |