Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605133 MIM: 612845 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCBP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NCBP2 - nuclear cap binding protein subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042540.1 | 1588 | UTR 3 | NP_001036005.1 | |||
NM_001308036.1 | 1588 | UTR 3 | NP_001294965.1 | |||
NM_007362.3 | 1588 | UTR 3 | NP_031388.2 | |||
XM_011512556.2 | 1588 | UTR 3 | XP_011510858.1 | |||
XM_011512557.2 | 1588 | UTR 3 | XP_011510859.1 | |||
XM_011512558.2 | 1588 | UTR 3 | XP_011510860.1 |
NCBP2-AS2 - NCBP2 antisense RNA 2 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SENP5 - SUMO1/sentrin specific peptidase 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |