Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAACTAGTCAAGATAAGTCTTTT[A/T]AAGTTATAAAAGTTTAATTTGATCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608307 MIM: 604155 | ||||||||||||||||||||
Literature Links: |
CPS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CPS1 - carbamoyl-phosphate synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122633.2 | Intron | NP_001116105.1 | ||||
NM_001122634.3 | Intron | NP_001116106.1 | ||||
NM_001875.4 | Intron | NP_001866.2 | ||||
XM_011510640.1 | Intron | XP_011508942.1 | ||||
XM_011510641.2 | Intron | XP_011508943.1 | ||||
XM_011510642.2 | Intron | XP_011508944.1 | ||||
XM_011510643.2 | Intron | XP_011508945.1 | ||||
XM_011510644.2 | Intron | XP_011508946.1 |
LANCL1 - LanC like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |