Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601262 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF16 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF16 - zinc finger protein 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029976.2 | 2112 | Silent Mutation | NP_001025147.2 | |||
NM_006958.2 | 2112 | Silent Mutation | NP_008889.2 | |||
XM_005272341.2 | 2112 | Silent Mutation | XP_005272398.1 | |||
XM_005272342.2 | 2112 | Silent Mutation | XP_005272399.1 | |||
XM_011517298.1 | 2112 | Silent Mutation | XP_011515600.1 |
Set Membership: |
HapMap |