Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGACTAGCTGTGAGGGGTATTTTA[A/G]CAAAATAGAAGTTTAGATCAAAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607680 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RCHY1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RCHY1 - ring finger and CHY zinc finger domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009922.2 | 2936 | UTR 3 | NP_001009922.1 | |||
NM_001278536.1 | 2936 | UTR 3 | NP_001265465.1 | |||
NM_001278537.1 | 2936 | UTR 3 | NP_001265466.1 | |||
NM_001278538.1 | 2936 | UTR 3 | NP_001265467.1 | |||
NM_001278539.1 | 2936 | UTR 3 | NP_001265468.1 | |||
NM_015436.3 | 2936 | UTR 3 | NP_056251.2 | |||
XM_011531838.2 | 2936 | UTR 3 | XP_011530140.1 | |||
XM_011531839.2 | 2936 | UTR 3 | XP_011530141.1 |