Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602026 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHYH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PHYH - phytanoyl-CoA 2-hydroxylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037537.1 | Intron | NP_001032626.1 | ||||
NM_001323080.1 | Intron | NP_001310009.1 | ||||
NM_001323082.1 | Intron | NP_001310011.1 | ||||
NM_001323083.1 | Intron | NP_001310012.1 | ||||
NM_001323084.1 | Intron | NP_001310013.1 | ||||
NM_006214.3 | Intron | NP_006205.1 |
Set Membership: |
HapMap |