Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 126065 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP24A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CYP24A1 - cytochrome P450 family 24 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000782.4 | 2065 | UTR 3 | NP_000773.2 | |||
NM_001128915.1 | 2065 | UTR 3 | NP_001122387.1 | |||
XM_005260304.4 | 2065 | UTR 3 | XP_005260361.1 | |||
XM_017027691.1 | 2065 | UTR 3 | XP_016883180.1 | |||
XM_017027692.1 | 2065 | Intron | XP_016883181.1 | |||
XM_017027693.1 | 2065 | UTR 3 | XP_016883182.1 |
Set Membership: |
HapMap DME Validated Inventoried |