Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGATATAGAACCAAAATTGTCGGG[C/G]GCGACCTCAGGGGGTTATTTCATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608034 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASPA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ASPA - aspartoacylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000049.2 | Intron | NP_000040.1 | ||||
NM_001128085.1 | Intron | NP_001121557.1 | ||||
XM_017024661.1 | Intron | XP_016880150.1 |
SPATA22 - spermatogenesis associated 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170695.1 | Intron | NP_001164166.1 | ||||
NM_001170696.1 | Intron | NP_001164167.1 | ||||
NM_001170697.1 | Intron | NP_001164168.1 | ||||
NM_001170698.1 | Intron | NP_001164169.1 | ||||
NM_001170699.1 | Intron | NP_001164170.1 | ||||
NM_001321336.1 | Intron | NP_001308265.1 | ||||
NM_001321337.1 | Intron | NP_001308266.1 | ||||
NM_032598.4 | Intron | NP_115987.2 |