Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC33 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC33 - coiled-coil domain containing 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287181.1 | Intron | NP_001274110.1 | ||||
NM_025055.4 | Intron | NP_079331.3 | ||||
NM_182791.3 | Intron | NP_877592.2 | ||||
XM_005254692.1 | Intron | XP_005254749.1 | ||||
XM_011522085.2 | Intron | XP_011520387.1 | ||||
XM_011522086.1 | Intron | XP_011520388.1 | ||||
XM_011522087.2 | Intron | XP_011520389.1 | ||||
XM_011522088.1 | Intron | XP_011520390.1 | ||||
XM_011522089.2 | Intron | XP_011520391.1 | ||||
XM_011522090.2 | Intron | XP_011520392.2 | ||||
XM_017022623.1 | Intron | XP_016878112.1 | ||||
XM_017022624.1 | Intron | XP_016878113.1 | ||||
XM_017022625.1 | Intron | XP_016878114.1 | ||||
XM_017022626.1 | Intron | XP_016878115.1 | ||||
XM_017022627.1 | Intron | XP_016878116.1 | ||||
XM_017022628.1 | Intron | XP_016878117.1 | ||||
XM_017022629.1 | Intron | XP_016878118.1 | ||||
XM_017022630.1 | Intron | XP_016878119.1 | ||||
XM_017022631.1 | Intron | XP_016878120.1 | ||||
XM_017022632.1 | Intron | XP_016878121.1 | ||||
XM_017022633.1 | Intron | XP_016878122.1 |