Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606647 | ||||||||||||||||||||||||||||||||
Literature Links: |
ARAP3 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
ARAP3 - ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022481.5 | Intron | NP_071926.4 | ||||
XM_005268497.1 | Intron | XP_005268554.1 | ||||
XM_005268498.1 | Intron | XP_005268555.1 | ||||
XM_005268499.1 | Intron | XP_005268556.1 | ||||
XM_005268500.3 | Intron | XP_005268557.1 | ||||
XM_006714792.1 | Intron | XP_006714855.1 | ||||
XM_011537677.2 | Intron | XP_011535979.1 |
FCHSD1 - FCH and double SH3 domains 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |