Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGAGGGCCAGGTGCCCCTTGGTT[C/T]GGGAGATGAAGAACAGCCTCCCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606881 MIM: 600477 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FHOD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FHOD1 - formin homology 2 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318202.1 | 783 | Intron | NP_001305131.1 | |||
NM_013241.2 | 783 | Intron | NP_037373.2 | |||
XM_006721180.1 | 783 | Intron | XP_006721243.1 | |||
XM_011523043.2 | 783 | Intron | XP_011521345.1 | |||
XM_011523044.1 | 783 | Intron | XP_011521346.1 | |||
XM_011523045.2 | 783 | Intron | XP_011521347.1 |
SLC9A5 - solute carrier family 9 member A5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323971.1 | 783 | Intron | NP_001310900.1 | |||
NM_001323972.1 | 783 | Intron | NP_001310901.1 | |||
NM_001323973.1 | 783 | Intron | NP_001310902.1 | |||
NM_001323974.1 | 783 | Intron | NP_001310903.1 | |||
NM_001323975.1 | 783 | Intron | NP_001310904.1 | |||
NM_004594.2 | 783 | Intron | NP_004585.1 | |||
XM_017023594.1 | 783 | UTR 5 | XP_016879083.1 |
Set Membership: |
HapMap |