Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602547 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR548P PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR548P - microRNA 548p | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ST8SIA4 - ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005668.5 | 3184 | UTR 3 | NP_005659.1 | |||
NM_175052.2 | 3184 | Intron | NP_778222.1 | |||
XM_005272078.2 | 3184 | Intron | XP_005272135.1 | |||
XM_011543630.1 | 3184 | UTR 3 | XP_011541932.1 |
Set Membership: |
HapMap |