Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF780A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ZNF780A - zinc finger protein 780A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010880.2 | 5848 | Intron | NP_001010880.2 | |||
NM_001142577.1 | 5848 | Intron | NP_001136049.1 | |||
NM_001142578.1 | 5848 | Intron | NP_001136050.1 | |||
NM_001142579.1 | 5848 | UTR 3 | NP_001136051.1 | |||
XM_005258773.2 | 5848 | UTR 3 | XP_005258830.1 | |||
XM_006723150.3 | 5848 | UTR 3 | XP_006723213.1 | |||
XM_011526771.2 | 5848 | UTR 3 | XP_011525073.1 | |||
XM_011526772.2 | 5848 | UTR 3 | XP_011525074.1 | |||
XM_017026611.1 | 5848 | UTR 3 | XP_016882100.1 | |||
XM_017026612.1 | 5848 | UTR 3 | XP_016882101.1 | |||
XM_017026613.1 | 5848 | UTR 3 | XP_016882102.1 | |||
XM_017026614.1 | 5848 | UTR 3 | XP_016882103.1 | |||
XM_017026615.1 | 5848 | UTR 3 | XP_016882104.1 | |||
XM_017026616.1 | 5848 | UTR 3 | XP_016882105.1 | |||
XM_017026617.1 | 5848 | UTR 3 | XP_016882106.1 | |||
XM_017026618.1 | 5848 | UTR 3 | XP_016882107.1 | |||
XM_017026619.1 | 5848 | UTR 3 | XP_016882108.1 |
Set Membership: |
HapMap Validated |