Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATATACTTAACAGAGCTAAGTGGT[C/T]ACCTTTACTTCACTCATATTCAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602778 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR181A2HG PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MIR181A2HG - MIR181A2 host gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NR6A1 - nuclear receptor subfamily 6 group A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278546.1 | Intron | NP_001265475.1 | ||||
NM_001489.4 | Intron | NP_001480.3 | ||||
NM_033334.3 | Intron | NP_201591.2 | ||||
XM_005251917.4 | Intron | XP_005251974.1 | ||||
XM_005251918.4 | Intron | XP_005251975.1 |