Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191350 MIM: 601772 MIM: 609806 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPAGT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DPAGT1 - dolichyl-phosphate N-acetylglucosaminephosphotransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001382.3 | 1815 | Missense Mutation | ATC,GTC | I,V 393 | NP_001373.2 | |
XM_005271422.2 | 1815 | Missense Mutation | ATC,GTC | I,V 419 | XP_005271479.1 | |
XM_011542648.1 | 1815 | Missense Mutation | ATC,GTC | I,V 312 | XP_011540950.1 | |
XM_017017293.1 | 1815 | Missense Mutation | ATC,GTC | I,V 286 | XP_016872782.1 | |
XM_017017294.1 | 1815 | UTR 3 | XP_016872783.1 | |||
XM_017017295.1 | 1815 | Missense Mutation | ATC,GTC | I,V 221 | XP_016872784.1 |
H2AFX - H2A histone family member X | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HMBS - hydroxymethylbilane synthase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |