Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTAGGCAGATGTCATTTCAGAGA[A/T]GAGGAAGCAAGTTCAGAACGGCTTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606239 MIM: 179514 MIM: 606887 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IKZF4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IKZF4 - IKAROS family zinc finger 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105369781 - uncharacterized LOC105369781 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB5B - RAB5B, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SUOX - sulfite oxidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000456.2 | Intron | NP_000447.2 | ||||
NM_001032386.1 | Intron | NP_001027558.1 | ||||
NM_001032387.1 | Intron | NP_001027559.1 | ||||
XM_005269112.1 | Intron | XP_005269169.1 | ||||
XM_017019905.1 | Intron | XP_016875394.1 | ||||
XM_017019906.1 | Intron | XP_016875395.1 | ||||
XM_017019907.1 | Intron | XP_016875396.1 | ||||
XM_017019908.1 | Intron | XP_016875397.1 |
Set Membership: |
HapMap |