Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTATGAACATTGTAAGTTACTCTG[C/G]GGAAAAACTCTATAAAACTAAAAGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601980 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIPF PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LIPF - lipase F, gastric type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198828.1 | Intron | NP_001185757.1 | ||||
NM_001198829.1 | Intron | NP_001185758.1 | ||||
NM_001198830.1 | Intron | NP_001185759.1 | ||||
NM_004190.3 | Intron | NP_004181.1 | ||||
XM_011540311.1 | Intron | XP_011538613.1 |
Set Membership: |
HapMap |