Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612054 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNOT9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CNOT9 - CCR4-NOT transcription complex subunit 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271634.1 | 54 | UTR 5 | NP_001258563.1 | |||
NM_001271635.1 | 54 | UTR 5 | NP_001258564.1 | |||
NM_005444.2 | 54 | UTR 5 | NP_005435.1 | |||
XM_011512138.2 | 54 | UTR 5 | XP_011510440.1 | |||
XM_017005248.1 | 54 | Intron | XP_016860737.1 | |||
XM_017005249.1 | 54 | UTR 5 | XP_016860738.1 |
USP37 - ubiquitin specific peptidase 37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020935.2 | 54 | Intron | NP_065986.2 | |||
XM_005246720.2 | 54 | Intron | XP_005246777.1 | |||
XM_005246721.3 | 54 | Intron | XP_005246778.1 | |||
XM_005246722.3 | 54 | Intron | XP_005246779.1 | |||
XM_005246724.1 | 54 | Intron | XP_005246781.1 | |||
XM_011511538.2 | 54 | Intron | XP_011509840.1 | |||
XM_011511541.2 | 54 | Intron | XP_011509843.1 | |||
XM_017004577.1 | 54 | Intron | XP_016860066.1 |
Set Membership: |
HapMap |