Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606907 MIM: 142590 MIM: 115441 MIM: 142610 MIM: 610433 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOM PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
APOM - apolipoprotein M | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BAG6 - BCL2 associated athanogene 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C6orf47 - chromosome 6 open reading frame 47 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CSNK2B - casein kinase 2 beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPANK1 - G-patch domain and ankyrin repeats 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199237.1 | 1534 | Silent Mutation | AGA,AGG | R,R 291 | NP_001186166.1 | |
NM_001199238.1 | 1534 | Silent Mutation | AGA,AGG | R,R 291 | NP_001186167.1 | |
NM_001199239.1 | 1534 | Silent Mutation | AGA,AGG | R,R 291 | NP_001186168.1 | |
NM_001199240.1 | 1534 | Silent Mutation | AGA,AGG | R,R 291 | NP_001186169.1 | |
NM_033177.3 | 1534 | Silent Mutation | AGA,AGG | R,R 291 | NP_149417.1 | |
XM_005249403.3 | 1534 | Silent Mutation | AGA,AGG | R,R 291 | XP_005249460.1 | |
XM_006715204.1 | 1534 | Silent Mutation | AGA,AGG | R,R 291 | XP_006715267.1 | |
XM_011514909.1 | 1534 | Silent Mutation | AGA,AGG | R,R 291 | XP_011513211.1 | |
XM_011514910.1 | 1534 | Silent Mutation | AGA,AGG | R,R 291 | XP_011513212.1 |
LY6G5B - lymphocyte antigen 6 complex, locus G5B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |