Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147678 MIM: 602665 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CASP1 - caspase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001223.4 | Intron | NP_001214.1 | ||||
NM_001257118.2 | Intron | NP_001244047.1 | ||||
NM_001257119.2 | Intron | NP_001244048.1 | ||||
NM_033292.3 | Intron | NP_150634.1 | ||||
NM_033293.3 | Intron | NP_150635.1 | ||||
NM_033294.3 | Intron | NP_150636.1 | ||||
NM_033295.3 | Intron | NP_150637.1 | ||||
XM_017018393.1 | Intron | XP_016873882.1 | ||||
XM_017018394.1 | Intron | XP_016873883.1 | ||||
XM_017018395.1 | Intron | XP_016873884.1 | ||||
XM_017018396.1 | Intron | XP_016873885.1 |
CASP5 - caspase 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |