Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGACAATAGGTACAGCAAAGAGAC[A/G]AATCAAGACTTGGGTCTCACAGAAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138245 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRIK1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GRIK1 - glutamate ionotropic receptor kainate type subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000830.4 | Intron | NP_000821.1 | ||||
NM_001320616.1 | Intron | NP_001307545.1 | ||||
NM_001320618.1 | Intron | NP_001307547.1 | ||||
NM_001320621.1 | Intron | NP_001307550.1 | ||||
NM_001320630.1 | Intron | NP_001307559.1 | ||||
NM_175611.2 | Intron | NP_783300.1 | ||||
XM_005260942.1 | Intron | XP_005260999.1 | ||||
XM_005260944.3 | Intron | XP_005261001.1 | ||||
XM_006723991.1 | Intron | XP_006724054.1 |
GRIK1-AS2 - GRIK1 antisense RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |