Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTATCCAAATTTAAGCATCTGA[C/T]GTGTGATATGAGTATATAACATATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604505 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIP11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
TRIP11 - thyroid hormone receptor interactor 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321851.1 | Intron | NP_001308780.1 | ||||
NM_004239.4 | Intron | NP_004230.2 | ||||
XM_017021787.1 | Intron | XP_016877276.1 | ||||
XM_017021788.1 | Intron | XP_016877277.1 |
Set Membership: |
HapMap |