Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614014 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNASE9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RNASE9 - ribonuclease A family member 9 (inactive) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001673.3 | 784 | UTR 3 | NP_001001673.2 | |||
NM_001110356.1 | 784 | UTR 3 | NP_001103826.1 | |||
NM_001110357.1 | 784 | UTR 3 | NP_001103827.1 | |||
NM_001110358.1 | 784 | UTR 3 | NP_001103828.1 | |||
NM_001110359.1 | 784 | UTR 3 | NP_001103829.1 | |||
NM_001110360.1 | 784 | UTR 3 | NP_001103830.1 | |||
NM_001110361.1 | 784 | UTR 3 | NP_001103831.1 | |||
NM_001289110.1 | 784 | UTR 3 | NP_001276039.1 |
Set Membership: |
HapMap |