Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCACGGCTGGCTTTCGCAGAGC[A/G]GCGAAGCTGATCTTGCTTTCTGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604572 MIM: 610057 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DNAJB1 - DnaJ heat shock protein family (Hsp40) member B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300914.1 | Intron | NP_001287843.1 | ||||
NM_001313964.1 | Intron | NP_001300893.1 | ||||
NM_006145.2 | Intron | NP_006136.1 | ||||
XM_006722733.1 | Intron | XP_006722796.1 | ||||
XM_006722734.3 | Intron | XP_006722797.1 | ||||
XM_011527956.2 | Intron | XP_011526258.1 |
MIR639 - microRNA 639 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TECR - trans-2,3-enoyl-CoA reductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321170.1 | Intron | NP_001308099.1 | ||||
NM_138501.5 | Intron | NP_612510.1 | ||||
XM_006722945.1 | Intron | XP_006723008.1 | ||||
XM_011528442.2 | Intron | XP_011526744.1 |
Set Membership: |
Validated |