Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATCCCTAAACTACCTGAGATTAC[A/G]GGCAAACATTTTCATGTACGTGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605296 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRWD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BRWD1 - bromodomain and WD repeat domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSMG1 - proteasome assembly chaperone 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261824.1 | Intron | NP_001248753.1 | ||||
NM_001320795.1 | Intron | NP_001307724.1 | ||||
NM_003720.3 | Intron | NP_003711.1 | ||||
NM_203433.2 | Intron | NP_982257.1 | ||||
XM_017028488.1 | Intron | XP_016883977.1 |
Set Membership: |
HapMap |