Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600023 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDH11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CDH11 - cadherin 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308392.1 | 3634 | UTR 3 | NP_001295321.1 | |||
NM_001797.3 | 3634 | UTR 3 | NP_001788.2 | |||
XM_005255761.3 | 3634 | UTR 3 | XP_005255818.1 | |||
XM_005255762.2 | 3634 | UTR 3 | XP_005255819.1 | |||
XM_005255763.2 | 3634 | UTR 3 | XP_005255820.1 | |||
XM_011522803.2 | 3634 | UTR 3 | XP_011521105.1 | |||
XM_017022830.1 | 3634 | UTR 3 | XP_016878319.1 |
Set Membership: |
HapMap |