Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608269 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC28A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC28A3 - solute carrier family 28 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199633.1 | 6442 | Intron | NP_001186562.1 | |||
NM_022127.2 | 6442 | Intron | NP_071410.1 | |||
XM_011518905.2 | 6442 | UTR 3 | XP_011517207.1 | |||
XM_011518906.2 | 6442 | UTR 3 | XP_011517208.1 | |||
XM_011518907.2 | 6442 | UTR 3 | XP_011517209.1 | |||
XM_011518908.2 | 6442 | UTR 3 | XP_011517210.1 | |||
XM_011518909.2 | 6442 | Intron | XP_011517211.1 | |||
XM_011518910.2 | 6442 | Intron | XP_011517212.1 |
Set Membership: |
HapMap |