Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614903 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNX16 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SNX16 - sorting nexin 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022133.3 | 2875 | UTR 3 | NP_071416.2 | |||
NM_152836.2 | 2875 | UTR 3 | NP_690049.1 | |||
NM_152837.2 | 2875 | UTR 3 | NP_690050.1 | |||
XM_005251282.4 | 2875 | UTR 3 | XP_005251339.1 | |||
XM_005251283.2 | 2875 | UTR 3 | XP_005251340.1 | |||
XM_011517574.2 | 2875 | UTR 3 | XP_011515876.1 |