Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
26 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611758 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OTUD3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
OTUD3 - OTU deubiquitinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015207.1 | 621 | Intron | NP_056022.1 | |||
XM_005245792.3 | 621 | Missense Mutation | CCC,CGC | P,R 15 | XP_005245849.1 | |
XM_005245793.3 | 621 | Missense Mutation | CCC,CGC | P,R 15 | XP_005245850.1 | |
XM_005245794.3 | 621 | Intron | XP_005245851.1 |
Set Membership: |
HapMap |