Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYO15B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MYO15B - myosin XVB | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001309242.1 | 2397 | Silent Mutation | CGA,CGG | R,R 846 | NP_001296171.1 | |
XM_017025120.1 | 2397 | Silent Mutation | CGA,CGG | R,R 628 | XP_016880609.1 | |
XM_017025121.1 | 2397 | Silent Mutation | CGA,CGG | R,R 623 | XP_016880610.1 | |
XM_017025122.1 | 2397 | Silent Mutation | CGA,CGG | R,R 628 | XP_016880611.1 | |
XM_017025123.1 | 2397 | Silent Mutation | CGA,CGG | R,R 628 | XP_016880612.1 | |
XM_017025124.1 | 2397 | Silent Mutation | CGA,CGG | R,R 628 | XP_016880613.1 | |
XM_017025125.1 | 2397 | Silent Mutation | CGA,CGG | R,R 628 | XP_016880614.1 | |
XM_017025126.1 | 2397 | Silent Mutation | CGA,CGG | R,R 628 | XP_016880615.1 | |
XM_017025127.1 | 2397 | Silent Mutation | CGA,CGG | R,R 399 | XP_016880616.1 | |
XM_017025128.1 | 2397 | Silent Mutation | CGA,CGG | R,R 390 | XP_016880617.1 | |
XM_017025129.1 | 2397 | Silent Mutation | CGA,CGG | R,R 107 | XP_016880618.1 | |
XM_017025130.1 | 2397 | Silent Mutation | CGA,CGG | R,R 99 | XP_016880619.1 | |
XM_017025131.1 | 2397 | Silent Mutation | CGA,CGG | R,R 64 | XP_016880620.1 | |
XM_017025132.1 | 2397 | UTR 5 | XP_016880621.1 | |||
XM_017025133.1 | 2397 | UTR 5 | XP_016880622.1 | |||
XM_017025134.1 | 2397 | UTR 5 | XP_016880623.1 | |||
XM_017025135.1 | 2397 | Intron | XP_016880624.1 | |||
XM_017025136.1 | 2397 | Intron | XP_016880625.1 | |||
XM_017025137.1 | 2397 | Intron | XP_016880626.1 | |||
XM_017025138.1 | 2397 | Intron | XP_016880627.1 | |||
XM_017025139.1 | 2397 | Intron | XP_016880628.1 | |||
XM_017025140.1 | 2397 | Intron | XP_016880629.1 | |||
XM_017025141.1 | 2397 | Silent Mutation | CGA,CGG | R,R 628 | XP_016880630.1 | |
XM_017025142.1 | 2397 | Silent Mutation | CGA,CGG | R,R 628 | XP_016880631.1 | |
XM_017025143.1 | 2397 | Intron | XP_016880632.1 | |||
XM_017025144.1 | 2397 | UTR 5 | XP_016880633.1 |
Set Membership: |
HapMap |