Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616062 MIM: 602581 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKLE2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANKLE2 - ankyrin repeat and LEM domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GOLGA3 - golgin A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172557.1 | 6543 | Intron | NP_001166028.1 | |||
NM_005895.3 | 6543 | UTR 3 | NP_005886.2 | |||
XM_005266164.4 | 6543 | UTR 3 | XP_005266221.1 | |||
XM_005266167.4 | 6543 | UTR 3 | XP_005266224.1 | |||
XM_006719736.3 | 6543 | UTR 3 | XP_006719799.1 | |||
XM_006719737.3 | 6543 | UTR 3 | XP_006719800.1 | |||
XM_011534793.2 | 6543 | UTR 3 | XP_011533095.1 | |||
XM_011534794.2 | 6543 | UTR 3 | XP_011533096.1 |
Set Membership: |
HapMap |