Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600805 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LAMA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LAMA3 - laminin subunit alpha 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000227.4 | Intron | NP_000218.3 | ||||
NM_001127717.2 | Intron | NP_001121189.2 | ||||
NM_001127718.2 | Intron | NP_001121190.2 | ||||
NM_001302996.1 | Intron | NP_001289925.1 | ||||
NM_198129.2 | Intron | NP_937762.2 | ||||
XM_011525978.2 | Intron | XP_011524280.1 | ||||
XM_011525979.2 | Intron | XP_011524281.1 | ||||
XM_011525980.2 | Intron | XP_011524282.1 | ||||
XM_011525981.2 | Intron | XP_011524283.1 | ||||
XM_011525982.2 | Intron | XP_011524284.1 | ||||
XM_017025743.1 | Intron | XP_016881232.1 | ||||
XM_017025744.1 | Intron | XP_016881233.1 |