Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601436 | ||||||||||||||||||||
Literature Links: |
LMBRD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LMBRD2 - LMBR1 domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007527.1 | 254 | Intron | NP_001007528.1 |
MIR580 - microRNA 580 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SKP2 - S-phase kinase-associated protein 2, E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243120.1 | 254 | UTR 5 | NP_001230049.1 | |||
NM_005983.3 | 254 | Silent Mutation | CTG,TTG | L,L 12 | NP_005974.2 | |
NM_032637.3 | 254 | Silent Mutation | CTG,TTG | L,L 12 | NP_116026.1 |