Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138470 MIM: 605996 MIM: 154040 MIM: 600478 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFB - complement factor B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DXO - decapping exoribonuclease | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR1236 - microRNA 1236 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NELFE - negative elongation factor complex member E | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SKIV2L - Ski2 like RNA helicase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006929.4 | 773 | Silent Mutation | GCC,GCT | A,A 128 | NP_008860.4 | |
XM_011514815.2 | 773 | Silent Mutation | GCC,GCT | A,A 128 | XP_011513117.1 |