Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGCCAGAGTGATGCCTCAGGCAC[C/G]GCTAGAAACCCAGGGCCGGACAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609948 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107986762 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC107986762 - uncharacterized LOC107986762 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF216 - ring finger protein 216 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207111.3 | 3850 | UTR 3 | NP_996994.1 | |||
NM_207116.2 | 3850 | UTR 3 | NP_996999.1 | |||
XM_005249785.2 | 3850 | UTR 3 | XP_005249842.1 | |||
XM_011515436.1 | 3850 | UTR 3 | XP_011513738.1 | |||
XM_017012363.1 | 3850 | UTR 3 | XP_016867852.1 | |||
XM_017012364.1 | 3850 | Intron | XP_016867853.1 | |||
XM_017012365.1 | 3850 | UTR 3 | XP_016867854.1 |
Set Membership: |
HapMap |