Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605397 | ||||||||||||||||||||
Literature Links: |
CD226 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD226 - CD226 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303618.1 | 4649 | Intron | NP_001290547.1 | |||
NM_001303619.1 | 4649 | Intron | NP_001290548.1 | |||
NM_006566.3 | 4649 | Intron | NP_006557.2 | |||
XM_005266642.3 | 4649 | UTR 3 | XP_005266699.1 | |||
XM_005266643.3 | 4649 | UTR 3 | XP_005266700.1 | |||
XM_006722374.3 | 4649 | UTR 3 | XP_006722437.1 | |||
XM_017025524.1 | 4649 | UTR 3 | XP_016881013.1 | |||
XM_017025525.1 | 4649 | UTR 3 | XP_016881014.1 | |||
XM_017025526.1 | 4649 | UTR 3 | XP_016881015.1 | |||
XM_017025527.1 | 4649 | UTR 3 | XP_016881016.1 |