Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609359 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HS1BP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HS1BP3 - HCLS1 binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022460.3 | 2294 | UTR 3 | NP_071905.3 | |||
XM_017004696.1 | 2294 | Intron | XP_016860185.1 | |||
XM_017004697.1 | 2294 | Intron | XP_016860186.1 | |||
XM_017004698.1 | 2294 | Intron | XP_016860187.1 | |||
XM_017004699.1 | 2294 | Intron | XP_016860188.1 | |||
XM_017004700.1 | 2294 | UTR 3 | XP_016860189.1 | |||
XM_017004701.1 | 2294 | Intron | XP_016860190.1 |