Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1QTNF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C1QTNF2 - C1q and tumor necrosis factor related protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031908.4 | 1148 | UTR 3 | NP_114114.2 | |||
XM_011534425.2 | 1148 | UTR 3 | XP_011532727.1 | |||
XM_011534426.2 | 1148 | UTR 3 | XP_011532728.1 | |||
XM_011534427.2 | 1148 | UTR 3 | XP_011532729.1 | |||
XM_017009013.1 | 1148 | UTR 3 | XP_016864502.1 |
CCNJL - cyclin J like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |